Learn More
Medchemexpress LLC VO-Ohpic trihydrate | 476310-60-8 | 98.0% | 50 MG
SDP
Supplier:  Medchemexpress LLC HY1307450MG
Encompass_Preferred
VO-Ohpic trihydrate is a highly potent inhibitor of PTEN with an IC50 of 46±10 nM. This sterically demanding molecule significantly inhibits PTEN activity even at low nanomolar concentrations. It is recognized as a specific and potent PTEN inhibitor, and is considered the most potent inhibitor of PTEN lipid phosphatase activity.
  • Highly potent inhibitor of PTEN with an IC50 of 46±10 nM
  • Significantly inhibits PTEN activity at low nanomolar concentrations
  • Specific and potent PTEN inhibitor
  • Most potent inhibitor of PTEN lipid phosphatase activity (IC50 of 35 nM)
Catalog No. 50-003-38669