Learn More
Medchemexpress LLC Voacangine | 510-22-5 | 1 MG

Supplier: Medchemexpress LLC HYN75361MG
Voacangine is an antagonist for TRPV1 and TRPM8, and an agonist for TRPA1 (EC50=8 μM). It competitively blocks capsaicin binding to TRPV1 (IC50=50 μM) and inhibits menthol binding to TRPM8 (IC50=9 μM). It is isolated from the root bark of Voacanga africana.
- Antagonist for TRPV1 and TRPM8.
- Agonist for TRPA1 (EC50=8 μM).
- Competitively blocks capsaicin binding to TRPV1 (IC50=50 μM).
- Competitively inhibits the binding of menthol to TRPM8 (IC50=9 μM).
- Shows noncompetitive inhibition against icilin (IC50=7 μM).
- Selectively abrogates chemical agonist-induced TRPM8 activation.
- Does not affect cold-induced TRPM8 activation.
- Alkaloid isolated from the root bark of Voacanga africana.
- Purity: 98.78%.
- Appearance: Solid, white to off-white.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.