Learn More
Medchemexpress LLC Voacangine | 510-22-5 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN75365MG
Encompass_Preferred
Voacangine is an alkaloid isolated from the root bark of *Voacanga africana*. It acts as an antagonist for TRPV1 and TRPM8, and an agonist for TRPA1. It competitively blocks capsaicin binding to TRPV1 and inhibits menthol binding to TRPM8. Voacangine selectively abrogates chemical agonist-induced TRPM8 activation without affecting cold-induced activation.
  • Acts as an antagonist for TRPV1 and TRPM8
  • Functions as an agonist for TRPA1
  • Competitively blocks capsaicin binding to TRPV1
  • Inhibits menthol binding to TRPM8
  • Suppresses in vitro angiogenesis
Catalog No. 50-004-19535