Learn More
TARGETMOL CHEMICALS INC Voclosporin 1MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T172351MG
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Voclosporin (ISAtx-247) is a novel, orally available calcium-modulated phosphatase (CN; PP2B) inhibitor and immunosuppressant used for treating lupus nephritis. Purity 99.85%
Catalog No. 50-287-6880