Learn More
Ace Glass, Inc. Volumetric flask, class A, 200mL, blue lettering, with individual certificate, includes 14/23 polyethylene stopper, sp/2
SDP
Supplier:  Ace Glass, Inc. 712220
Encompass
Volumetric flask, class A, 200mL, blue lettering, with individual certificate, includes 14/23 polyethylene stopper, sp/2
Catalog No. 50-974-77