Learn More
Medchemexpress LLC Volanesorsen sodium | 1573402-50-2 | 98.5% | C230H301N63Na19O125P19S19 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY145727A1MG
Encompass_Preferred
Volanesorsen sodium is an antisense oligonucleotide inhibitor of apolipoprotein CIII (apo-CIII) mRNA. It works by reducing triglyceride levels and improving insulin resistance.
  • Reduces triglyceride levels
  • Improves insulin resistance
  • Being investigated for the treatment of hypertriglyceridemia
  • Being investigated for familial chylosiderosis syndrome
  • Being investigated for type 2 diabetes
  • Antisense oligonucleotide inhibitor of apolipoprotein CIII (apo-CIII) mRNA
Catalog No. 50-003-54159