Learn More
TARGETMOL CHEMICALS INC VOLASERTIB 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T601910MG
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Volasertib (BI 6727) (BI-6727) is a potent inhibitor of PLK1 (IC50 0.87 nM) inducing mitotic arrest and apoptosis. It also inhibits PLK2/PLK3 (IC50s 5/56 nM). purity: 99%
Catalog No. 50-257-9649