Learn More
Selleck Chemical LLC Volasertib (BI 6727) 10mM/1mL 755038-65-4
SDP
Supplier:  Selleck Chemical LLC S223510MM/1ML
Encompass_Preferred
Volasertib (BI 6727) is a highly potent Plk1 inhibitor with IC50 of 0.87 nM in a cell-free assay. It shows 6- and 65-fold greater selectivity against Plk2 and Plk3. Volasertib induces cell cycle arrest and apoptosis in various cancer cells. Phase 3. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC1291348