Learn More
Hach Company Volatile Acids TNTplus Vial Test (50-2,500 mg/L), 25 Tests
Supplier:  Hach Company TNT872
Encompass
Volatile Acids TNTplus chemistry and your Hach spectrophotometer are engineered to simplify water analysis for accurate results, everytime.For determination of Volatile Acids by the Esterification method. Simple test allows quick process control - No need for titrations or involved water bath digestion.
Catalog No. 50-199-8157