Learn More
Volu Sol Hematology Buffer (pH 6.8), 128 oz / 3.78 L, Volu-Sol™
SDP
Supplier:  Volu Sol VWB128
Encompass_Preferred
Phosphate Buffer, intended for use as a buffer in various Romanowsky type staining procedures (Wright, Wright-Giemsa, Giemsa, May Grunwald, Jenner, and Leishman)
Catalog No. NC9477487