Learn More
CHEMSCIENCE Volumetric Flask Class A, Wide Mouth, 50 ML,PP Stopper No - 13, Clear Glass, Tolerance - ± 0.05,CS/48
Supplier:  CHEMSCIENCE 129.203.0448CS
Encompass_Preferred
Flasks are calibrated 'To Contain' at Class A tolerances, per ASTM E288 specifications. Supplied with closed bottom PP stopper to fit in the ground neck.Wide mouth design facilitates mixing, pipette access, sampling and filling. Unique flat bottom interior permits the use of a standard stir bar.
Catalog No. NC1884061