Learn More
Chemglass Life Sciences 500mL Volumetric Flask, Class A, #19 Outer Stopper Neck, Mixing Bulb
SDP
Supplier:  Chemglass Life Sciences CG160107
Encompass_Preferred
500mL Volumetric Flask, Class A, #19 Outer Stopper Neck, Mixing Bulb. Similar to CG-1600 but with the addition of a mixing bulb between the calibration line and the stopper. The mixing bulb contains approximately one tenth of the flask capacity and aids in the mixing or dissolving of the flask contents. Supplied complete with one solid glass pennyhead stopper. For additional stoppers, see CG-3018.
Catalog No. CG160107