Learn More
Vee Gee Volumetric Pipet, 8 mL
Supplier:  Vee Gee 2040A8
Encompass_Preferred
Volumetric Pipettes are meticulously manufactured for tolerances and accuracy in order to ensure pinpoint precision for all of your liquid handling needs. Volumetric pipettes are graduated to the tip and made out of high-quality borosilicate glass. These pipettes also have graduation markings and color-coded bands that are permanently enameled to the glass which allows for quick identification of different volume ranges. From micro to macro, our line of glass volumetric pipettes cover a wide range of volumes* Made out of high-quality borosilicate glass* Wide range of sizes to choose from* Reusable* Color-coded with permanent enamel markings* Calibrated To Deliver* Meets ASTM E-969 Class A Specifications & USP StandardsSPECIFICATIONS:* Color Code: Blue* Color: Clear* Length: 430 mm* Material: Borosilicate Glass* Meet ASTM E-969 Class A Specifications and USP Standards for volumetric glassware: 8.0 mL* Tolerance: ±0.02 mL* Quantity: Pack of 12
Catalog No. NC2386476