Learn More
Medchemexpress LLC Von willebrand factor antibody (YA2406)(PBS only) | 10 UL
SDP
Supplier:  Medchemexpress LLC HYP8266110UL
Encompass_Preferred
The von Willebrand Factor Antibody (YA2406) is a rabbit-derived, non-conjugated IgG monoclonal antibody that targets von Willebrand Factor. This antibody plays a critical role in hemostasis, mediating platelet adhesion to vascular injury sites and acting as a chaperone for coagulation factor VIII, stabilizing its structure and protecting it from premature clearance. It is suitable for research applications such as Western Blot and Immunohistochemistry-Paraffin.
  • Rabbit-derived host
  • Recombinant monoclonal antibody
  • IgG isotype
  • Non-conjugated
  • Reacts with rat species
  • Targets von Willebrand Factor
  • Affinity purified
  • Suitable for Western Blot (WB) and Immunohistochemistry-Paraffin (IHC-P) applications
  • Secreted subcellular localization
  • Expressed in plasma
Catalog No. 50-004-44612