Learn More
Medchemexpress LLC von Willebrand Factor Antibody (YA2406)(PBS only) | 50 UL
SDP
Supplier:  Medchemexpress LLC HYP8266150UL
Encompass_Preferred
This antibody is a rabbit-derived and non-conjugated IgG monoclonal antibody, specifically targeting von Willebrand Factor (vWF). vWF plays a critical role in hemostasis by mediating platelet adhesion to sites of vascular injury and acts as a carrier for coagulation factor VIII.
  • Rabbit-derived IgG monoclonal antibody
  • Non-conjugated
  • Targets von Willebrand Factor
  • Affinity purified
  • Supplied in PBS, pH 7.4
  • Stored at -20°C for 1 year, avoiding repeated freeze/thaw cycles
  • Liquid appearance
  • Suitable for WB and IHC-P applications
Catalog No. 50-004-44692