Learn More
Medchemexpress LLC Vonafexor | 1192171-69-9 | 99.38% | 489.76 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY10919710MM/1ML
Encompass_Preferred
Vonafexor (EYP001) is an orally active, non-steroidal and selective FXR agonist. It demonstrates significant HBsAg reduction when combined with Peg-IFNα and can be utilized for anti-HBV research. In vitro, Vonafexor inhibits the HBV replication cycle in HepaRG cells, leading to substantial reductions in HBV DNA, HBsAg, and HBeAg secretion, with additive effects when combined with Entecavir or Tenofovir.
  • Orally active, non-steroidal, and selective FXR agonist.
  • Shows significant HBsAg reduction when combined with Peg-IFNα.
  • Can be used for anti-HBV research.
  • Inhibits HBV replication cycle in HepaRG cells.
  • Additive anti-HBV effects when combined with Entecavir or Tenofovir.
Catalog No. 50-003-45281