Learn More
Medchemexpress LLC Vonafexor (EYP001) | 1192171-69-9 | 99.4% | 489.76 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY10919750MG
Encompass_Preferred
Vonafexor (EYP001) is an orally active, non-steroidal, and selective FXR agonist. It significantly reduces HBsAg when combined with Peg-IFNα and can be used for anti-HBV research. In vitro, Vonafexor inhibits HBV replication in HepaRG cells, leading to significant reductions in HBV DNA, HBsAg, and HBeAg secretion. This effect is enhanced when combined with Entecavir or Tenofovir.
  • Orally active, non-steroidal, and selective FXR agonist.
  • Shows significant HBsAg reduction when combined with Peg-IFNα.
  • Can be used for anti-HBV research.
  • Inhibits HBV replication cycle in HepaRG cells.
  • Additive effects when combined with Entecavir or Tenofovir in inhibiting HBV DNA, HBsAg, and HBeAg secretion.
Catalog No. 50-003-45282