Learn More
Cayman Chemical Vonpraznfumarate 5g
SDP
Supplier:  Cayman Chemical 242005 G
Encompass_Preferred
A selective and reversible proton pump inhibitor; inhibits gastric H+/K+ ATPase (IC50 = 17 nM); does not inhibit porcine Na+/K+ ATPase activity (10 µM); maintains its inhibitory effect in weakly acidic and neutral pH conditions (IC50s = 19 and 28 nM, respectively); inhibits histamine-stimulated acid secretion in a dose-dependent manner in rats (1, 2, and 4 mg/kg); inhibits acid secretion for more than 48 hours in dogs (0.1 to 1 mg/kg)
Catalog No. 50-293-3338