Learn More
Selleck Chemical LLC Voreloxin
SDP
Supplier:  Selleck Chemical LLC S75185MG
Encompass_Preferred
Voreloxin hydrochloride (SNS-595 Vosaroxin) is a potent Topoisomerase II inhibitor with broad-spectrum anti-tumor activity Phase 2
Catalog No. 50-000-04435