Learn More
Medchemexpress LLC Voreloxin hydrochloride | 175519-16-1 | 99.6% | 437.90 g/mol | C18H20ClN5O4S | 10 MG
SDP
Supplier:  Medchemexpress LLC HY1651810MG
Encompass_Preferred
Voreloxin hydrochloride is a research-grade anticancer compound and a first-in-class topoisomerase II inhibitor. It intercalates into DNA and induces site-selective double-strand breaks (DSB), causing G2 cell-cycle arrest and apoptosis. Supplied as a hydrochloride salt for laboratory research use only.
  • First-in-class mechanism: topoisomerase II inhibition through DNA intercalation.
  • Triggers site-selective DNA double-strand breaks and G2 arrest.
  • High purity suitable for biochemical and cell-based assays.
  • Well-characterized chemical identity: molecular formula C18H20ClN5O4S, molecular weight 437.90 g/mol.
  • Available in accurately weighed small pack sizes for precise dosing in experiments.
Catalog No. 50-002-89305