Learn More
Apexbio Technology LLC Voriconazole 137234-62-9 100mg
SDP
Supplier:  Apexbio Technology LLC A4320100
Encompass_Preferred
Voriconazole is a triazole antifungal agent functioning through the inhibition of lanosterol 14 -demethylase (CYP51) a cytochrome P450 enzyme responsible for fungal cell membrane ergosterol biosynthesis Voriconazole exhibits inhibitory activity at an IC50 of approximately 53 nM Its antifungal spectrum covers various fungal species including Candida species Aspergillus species Cryptococcus neoformans Trichosporon beigelii and Saccharomyces cerevisiae Due to these properties Voriconazole is frequently utilized in biomedical and pharmacological research involving fungal pathogenesis antifungal susceptibility studies and sterol biosynthesis pathway elucidation assays
Catalog No. 50-000-61467