Learn More
Medchemexpress LLC Voriconazole | 137234-62-9 | 99.5% | C16H14F3N5O | 5 MG
SDP
Supplier:  Medchemexpress LLC HY762005MG
Encompass_Preferred
Voriconazole (UK-109496) is a second-generation, broad-spectrum triazole antifungal agent. It inhibits fungal ergosterol biosynthesis by inhibiting 14-α-lanosterol demethylation, a process mediated by fungal cytochrome P450 enzymes. It is also capable of penetrating the blood-brain barrier (BBB).
  • Inhibits fungal ergosterol biosynthesis.
  • Inhibits 14-α-lanosterol demethylation.
  • Penetrates the blood-brain barrier.
  • Active against *S. apiospermum* (MIC 0.5 μg/mL) and *C. neoformans* (MIC 0.125-0.25 μg/mL).
  • Disrupts fungal cell membranes and halts growth.
Catalog No. 50-004-34188