Learn More
Medchemexpress LLC Voriconazole | 137234-62-9 | 99.89% | C16H14F3N5O | 1 ML
SDP
Supplier:  Medchemexpress LLC HY7620010MM/1ML
Encompass_Preferred
Voriconazole is a second-generation, broad-spectrum triazole antifungal agent that inhibits fungal ergosterol biosynthesis by inhibiting 14-α-lanosterol demethylation, a process mediated by fungal cytochrome P450 enzymes. It can penetrate the blood-brain barrier (BBB).
  • Inhibits fungal ergosterol biosynthesis
  • Penetrates the blood-brain barrier
  • Potent activity against S. apiospermum and C. neoformans
  • Inhibits cytochrome P450 (CYP)-dependent enzyme 14-alpha-sterol demethylase
Catalog No. 50-004-34593