Learn More
Medchemexpress LLC Voriconazole | 137234-62-9 | 99.9% | C16H14F3N5O | 50 MG
SDP
Supplier:  Medchemexpress LLC HY7620050MG
Encompass_Preferred
Voriconazole (UK-109496) is a second-generation, broad-spectrum triazole antifungal agent that inhibits fungal ergosterol biosynthesis. It exerts its antifungal activity by inhibition of 14-alpha-lanosterol demethylation, which is mediated by fungal cytochrome P450 enzymes. It can penetrate the blood brain barrier (BBB).
  • Second-generation, broad-spectrum triazole antifungal agent
  • Inhibits fungal ergosterol biosynthesis
  • Inhibits 14-alpha-lanosterol demethylation
  • Mediated by fungal cytochrome P450 enzymes
  • Penetrates the blood brain barrier (BBB)
  • Active against S. apiospermum and C. neoformans
  • Disrupts cell membrane and halts fungal growth
Catalog No. 50-004-34652