Learn More
Medchemexpress LLC Voriconazole | 137234-62-9 | 99.90% | C16H14F3N5O | 50 MG
SDP
Supplier:  Medchemexpress LLC HY76200R50MG
Encompass_Preferred
Voriconazole is an analytical standard and a second-generation, broad-spectrum triazole antifungal agent. It inhibits fungal ergosterol biosynthesis by blocking 14-α-lanosterol demethylation, a process mediated by fungal cytochrome P450 enzymes. This product is suitable for research and analytical applications.
  • Used as a reference standard supplied assay
  • Commonly used in qualitative, quantitative, and methodological research experiments
  • Suitable for HPLC, GC, and MS applications
Catalog No. 50-004-34653