Learn More
Medchemexpress LLC Voriconazole | 137234-62-9 | MFCD00905717 | 99.9% | 349.31 g/mol | C16H14F3N5O | 10 MG

Supplier: Medchemexpress LLC HY76200R10MG
Voriconazole analytical standard is a second-generation, broad-spectrum triazole antifungal provided as a high-purity reference material for research and analytical testing. It is intended for use in HPLC and other analytical assays to support quantitation and method validation.
- Analytical standard for research and analytical applications.
- High purity (99.9%).
- Molecular formula C16H14F3N5O.
- Molecular weight 349.31 g/mol.
- CAS number 137234-62-9.
- Supplied in small pack sizes suitable for analytical use, e.g., 10 mg.
- Antifungal mechanism: inhibits 14-α-lanosterol demethylation.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.