Learn More
Cayman Chemical VorIconazole-d3 500ug
SDP
Supplier:  Cayman Chemical 17803500 UG
Encompass_Preferred
An internal standard for the quantification of voriconazole by GC- or LC-MS
Catalog No. 50-296-6273