Learn More
Medchemexpress LLC Voriconazole Impurity 1 | 137330-52-0 | ≥98.0% | C16H14F3N5O | 25 MG

Supplier: Medchemexpress LLC 25MGHY76200A
Voriconazole Impurity 1 is a chemical impurity of Voriconazole (HY-76200). This product has a molecular weight of 349.31, a chemical formula of C16H14F3N5O, and a CAS Number of 137330-52-0. It appears as a solid and is suitable for research use only.
- White to off-white solid appearance
- Minimum purity of 98.0%
- Chemical formula C16H14F3N5O
- Molecular weight 349.31
- CAS number 137330-52-0
- Intended for research use only
- Store at 4°C, sealed, and away from moisture
- When in solvent, store at -80°C for 6 months or -20°C for 1 month, sealed and away from moisture
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.