Learn More
Cayman Chemical VorIconazole N-oxIde 1mg
SDP
Supplier:  Cayman Chemical 280471 MG
Encompass_Preferred
A major metabolite of voriconazole; formed via oxidation of voriconazole by CYP3A4 and CYP2C19
Catalog No. 50-293-8815