Learn More
Medchemexpress LLC Voriconazole N-oxide hydrochloride | 00-00-0 | MFCD09841413 | 99.3% | C16H15ClF3N5O2 | 10MG
SDP
Supplier:  Medchemexpress LLC HY137989A10MG
Encompass_Preferred
Voriconazole N-oxide hydrochloride is the hydrochloride salt of the major metabolite of the antifungal agent voriconazole. It is provided as a high-purity reference standard for research and analytical applications, including metabolite profiling, pharmacokinetic studies, and method validation. Follow supplier safety documentation and handling instructions due to reported phototoxicity and photocarcinogenicity in related compounds.
  • High purity suitable for analytical reference and standards.
  • Available in small milligram quantities for research use.
  • Suitable for metabolite identification and pharmacokinetic studies.
  • Useful for LC-MS and method development as a reference compound.
  • Provided with datasheet and handling instructions for safe use.
Catalog No. 50-002-59025