Learn More
Selleck Chemical LLC Vorinostat (SAHA) S1047-2g
SDP
Supplier:  Selleck Chemical LLC S10472G
Encompass_Preferred
Vorinostat (SAHA) is an HDAC inhibitor with IC50 of 10 nM in a cell-free assay Vorinostat abrogates productive HPV-18 DNA amplification
Catalog No. 50-311-1045