Learn More
Medchemexpress LLC Vorinostat-d5 1mg | 1132749-48-4 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY1154121MG
Encompass_Preferred
Vorinostat-d5 is the deuterium-labeled analogue of vorinostat (suberoylanilide hydroxamic acid) supplied as a stable-isotope internal standard for quantitative NMR and mass spectrometry. It is provided as a white to off-white solid with high purity suitable for analytical tracer use.
  • Deuterium-labeled internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • High purity suitable for analytical reference standards and tracer work.
  • Supplied in small, milligram quantities for method development and calibration.
  • Stable solid form with white to off-white appearance for easy handling and storage.
  • Chemical formula C14H15D5N2O3 and molecular weight 269.35 g/mol.
Catalog No. 50-002-29869