Learn More
Medchemexpress LLC HY-109019 5mg Medchemexpress, Vorolanib CAS:1013920-15-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1090195MG
Encompass_Preferred
Medchemexpress, HY-109019 5mg Vorolanib CAS:1013920-15-4 Vorolanib (CM082; X-82) is an orally active, multikinase VEGFR/PDGFR inhibitor. Vorolanib minimizes toxicity, disrupts tumor angiogenesis and tumor cell proliferation, and induces of tumor cell death. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8648