Learn More
Research Products International Corp Vortex-Genie Pulse, Digital Microplate
SDP
Supplier:  Research Products International Corp 155578
Encompass
Exclusive precise vortex orbit guarantees true vortexing action in each well of the microplate, at all times. Accurate speed and time control guarantee outstanding reliability and mixing consistency. MicroPlate Genies are ideal for applications such as small volume mixing, PCR Preparation, Digestions, as well as Cell Disruption.
Catalog No. 50-197-6987