Learn More
Thomas Scientific Horizontal head attachment for BenchMixer Vortexer; 2x50ml tubes
SDP
Supplier:  Thomas Scientific 1213F88
Encompass
Best Value! The Mortexer™ is MORE than your standard vortex mixer. The unique design of the Multi-Head™ (included) features a traditional cup head for general purpose mixing, plus holds up to eight microcentrifuge tubes. The drive system is balanced with Q-Drive™ technology, providing a smooth vortexing action, while minimizing noise and excessive vibration. This results in quieter, more efficient and longer lasting operation. Increased speed, up to 3400 rpm produces an instant vortex of even the largest sample sizes, including full 50ml tubes. Other features include: "no-walk" suction feet, cold room/incubator compatibility and spill-proof electronic controls. Two year warranty. Unique Multi-Head™ for general purpose vortexing PLUS holds up to eight microtubes (Patent Pending) Powerful motor for INSTANT vortexing Continuous or "touch" operation Q-Drive™ dynamic balancing system Variable speed from 200 to 3400 rpm Wide variety of access
Catalog No. 50-146-1254