Learn More
Medchemexpress LLC HY-15414 10mg Medchemexpress, Vortioxetine CAS:508233-74-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1541410MG
Encompass_Preferred
Medchemexpress, HY-15414 10mg Vortioxetine CAS:508233-74-7 Vortioxetine is a inhibitor of 5-HT1A, 5-HT1B, 5-HT3A, 5-HT7 receptor and SERT, with Ki values of 15 nM, 33 nM, 3.7 nM, 19 nM and 1.6 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1720