Learn More
Medchemexpress LLC Vortioxetine | 508233-74-7 | C18H22N2S | 1 G
SDP
Supplier:  Medchemexpress LLC HY154141G
Encompass_Preferred
Vortioxetine (Lu AA 21004) is a multimodal serotonergic agent that acts as an antagonist of 5-HT3A and 5-HT7 receptors and an inhibitor of the serotonin transporter (SERT). It also functions as a 5-HT1A agonist and a partial agonist of 5-HT1B.
  • Antagonist of 5-HT3A and 5-HT7 receptors
  • Inhibitor of serotonin transporter (SERT)
  • Agonist of 5-HT1A receptor
  • Partial agonist of 5-HT1B receptor
  • Intended for research use only
  • Does not cause cognitive or psychomotor impairment
Catalog No. 50-003-76652