Learn More
Medchemexpress LLC Vortioxetine | 508233-74-7 | C18H22N2S | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY15414R
Encompass_Preferred
Vortioxetine is an analytical standard and multimodal serotonergic agent. It functions as an antagonist of 5-HT3A and 5-HT7 receptors, an inhibitor of serotonin transporter (SERT), an agonist of 5-HT1A receptors, and a partial agonist of 5-HT1B receptors. This compound is intended for research and analytical applications, serving as a reference standard for various assay purposes.
  • Antagonist of 5-HT3A and 5-HT7 receptors
  • Inhibitor of serotonin transporter (SERT)
  • Agonist of 5-HT1A receptors
  • Partial agonist of 5-HT1B receptors
  • Used as an analytical reference standard
  • Suitable for qualitative, quantitative, and methodological research experiments (HPLC, GC, MS)
Catalog No. 50-003-75069