Learn More
Medchemexpress LLC Vortioxetine hydrobromide | 960203-27-4 | C18H23BrN2S | 1 ML
SDP
Supplier:  Medchemexpress LLC HY15414A10MM/1ML
Encompass_Preferred
Vortioxetine hydrobromide (Lu AA 21004) is a compound that acts as an antagonist of 5-HT3A and 5-HT7 receptors and an inhibitor of the 5-hydroxytryptamine transporter (SERT). It also functions as a 5-HT1A agonist and a partial 5-HT1B agonist.
  • Antagonist of 5-HT3A and 5-HT7 receptors
  • Inhibitor of 5-hydroxytryptamine transporter (SERT)
  • Agonist of 5-HT1A receptors
  • Partial agonist of 5-HT1B receptors
  • Increases cell proliferation and survival
  • Does not cause cognitive or psychomotor impairment
Catalog No. 50-003-76353