Learn More
Medchemexpress LLC Vortioxetine (hydrobromide) | 960203-27-4 | C18H23BrN2S | 200 MG
SDP
Supplier:  Medchemexpress LLC HY15414200MG
Encompass_Preferred
Vortioxetine hydrobromide is an antagonist of 5-HT3A and 5-HT7 receptors and an inhibitor of the 5-hydroxytryptamine transporter (SERT). It also acts as a 5-HT1A agonist and a partial 5-HT1B agonist. This compound is intended for research use only.
  • Antagonist of 5-HT3A and 5-HT7 receptors
  • Inhibitor of 5-hydroxytryptamine transporter (SERT)
  • 5-HT1A agonist
  • Partial 5-HT1B agonist
  • Intended for research use only
Catalog No. 50-003-76456