Learn More
Medchemexpress LLC Voruciclib | 1000023-04-0 | MFCD28386202 | 99.8% | 469.84 g/mol | C22H19ClF3NO5 | 10MM 1ML
SDP
Supplier:  Medchemexpress LLC HY1242210MM 1ML
Encompass_Preferred
Voruciclib is a selective cyclin-dependent kinase (CDK) inhibitor with potent activity against CDK9 and inhibitory activity against several other CDKs. Supplied as a 10 mM solution in DMSO (1 mL), it is intended for in vitro biochemical and cellular studies of transcriptional regulation, MCL-1 suppression, and apoptosis-related pathways.
  • Potent CDK9 inhibition in the low-nanomolar range.
  • Represses MCL-1 expression in cellular models.
  • Provided as a ready-to-use 10 mM DMSO solution (1 mL) for assays.
  • High purity (99.8%) suitable for research applications.
  • Documented biochemical potency and selectivity data.
  • Applicable to mechanistic and preclinical pathway studies.
Catalog No. 50-002-35363