Learn More
Medchemexpress LLC Voruciclib hydrochloride (2S,3R) | 2920469-94-7 | 98.0% | 506.30 | C22H20Cl2F3NO5 | 10MM 1ML
SDP
Supplier:  Medchemexpress LLC HY12422B10MM 1ML
Encompass_Preferred
(2S,3R)-Voruciclib hydrochloride is the (2S,3R) enantiomer of voruciclib, an orally active cyclin-dependent kinase (CDK) inhibitor used in research to probe cell-cycle regulation and CDK-related signaling. Supplied as the hydrochloride salt, it is provided as a solid powder and as a 10 mM solution in DMSO for assay use.
  • Enantiomeric hydrochloride salt for stereospecific studies.
  • High purity (≥98%) suitable for biochemical and cell-based assays.
  • Available as powder and 10 mM DMSO solution for immediate use.
  • Molecular weight 506.30 and CAS 2920469-94-7 for clear identification.
  • Intended for research on CDK-mediated cell-cycle and signaling pathways.
Catalog No. 50-002-38466