Learn More
Medchemexpress LLC Voruciclib hydrochloride (2S,3R) | 2920469-94-7 | 98.0% | 506.30 g·mol⁻¹ | C22H20Cl2F3NO5 | 10MG

Supplier: Medchemexpress LLC HY12422B10MG
(2S,3R)-Voruciclib hydrochloride is the enantiomer of voruciclib hydrochloride and an orally active cyclin-dependent kinase (CDK) inhibitor used in biochemical and cellular research. Supplied as the hydrochloride salt, it has CAS 2920469-94-7, molecular formula C22H20Cl2F3NO5, and molecular weight 506.30 g·mol⁻¹.
- Purity 98.0%.
- Hydrochloride salt and enantiomeric (2S,3R) form.
- Molecular weight 506.30 g·mol⁻¹; formula C22H20Cl2F3NO5.
- Intended for biochemical assays and cellular studies targeting CDKs.
- Available in milligram pack sizes, including 10 mg.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.