Learn More
Medchemexpress LLC HY-18681 25mg , Voxelotor CAS:1446321-46-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1868125MG
Encompass_Preferred
HY-18681 25mg Voxelotor CAS: 1446321-46-5 Voxelotor (GBT 440) is a potent inhibitor of haemoglobin S (HbS) polymerization. Voxelotor has the potential for sickle cell disease (SCD) treatment. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9559