Learn More
Medchemexpress LLC Voxilaprevir | 1535212-07-7 | 99.9% | 868.93 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY19840
Encompass_Preferred
Voxilaprevir is a noncovalent, reversible inhibitor of HCV NS3/4A protease with pangenotypic antiviral activity. It inhibits genotype 1b and 3a wild-type NS3 proteases with Ki values of 0.038 nM and 0.066 nM, respectively. It is an orally active direct-acting antiviral agent (DAA) and can be used for HCV infection research.
  • Noncovalent, reversible inhibitor of HCV NS3/4A protease.
  • Pangenotypic antiviral activity.
  • Inhibits genotype 1b and 3a wild-type NS3 proteases with Ki values of 0.038 nM and 0.066 nM, respectively.
  • Orally active direct-acting antiviral agent (DAA).
  • Used for HCV infection research.
  • Solid appearance.
  • White to off-white color.
Catalog No. 50-004-03168