Learn More
Medchemexpress LLC Voxvoganan trihydrochloride | 99.8% | 897.46 | C43H72Cl3N11O3 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY119123A5MG
Encompass_Preferred
Voxvoganan trihydrochloride is the trihydrochloride salt of voxvoganan (LTX-109), an investigational topical antimicrobial peptide that acts via membrane-lysing activity. Supplied as a solid research compound, it is intended for in vitro and preclinical research and is provided with supporting analytical and safety documentation.
  • High purity (99.8%) suitable for analytical and biological studies.
  • Solid form for accurate microgram-scale dosing.
  • Available in small-scale quantities: 5 MG, 10 MG, 25 MG, 50 MG, 100 MG.
  • Supplied with datasheet, certificate of analysis, H NMR, RP-HPLC, and MS files.
  • Molecular weight 897.46 and chemical formula C43H72Cl3N11O3.
Catalog No. 50-002-64593