Learn More
Medchemexpress LLC HY-122234 5mg , VPC-18005 CAS:2242480-48-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1222345MG
Encompass_Preferred
HY-122234 5mg VPC-18005 CAS: 2242480-48-2 VPC-18005 inhibits ERG-induced transcription and interacts directly with the ERG-ETS domain, and disrupts the ERG binding to DNA. VPC-18005 is a potent inhibitor of luciferase activity.
Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1698