Learn More
Medchemexpress LLC VPC-80051 racemate | 877969-69-2 | 99.0% | 301.29 g/mol | C16H13F2N3O | 10MG
SDP
Supplier:  Medchemexpress LLC HY12607610MG
Encompass_Preferred
VPC-80051 racemate is the racemic form of a small-molecule inhibitor of the hnRNP A1 splicing factor, supplied for laboratory research into RNA splicing and related cellular mechanisms. The compound has molecular formula C16H13F2N3O, molecular weight 301.29 g/mol, and CAS 877969-69-2; it is provided in small milligram quantities suitable for biochemical and cellular assays.
  • Available in 1 mg, 5 mg, and 10 mg pack sizes.
  • Approximately 99.0% purity.
  • Molecular formula C16H13F2N3O and molecular weight 301.29 g/mol.
  • Powder stable at -20°C for 3 years and 4°C for 2 years; in solvent stable at -80°C for 6 months and -20°C for 1 month.
  • Suitable for hnRNP A1 and RNA splicing assays and related cellular studies.
  • Supplied as a racemate for studies involving stereochemistry or isomeric activity.
Catalog No. 50-002-35547