Learn More
Cell Biolabs Inc QuickTiter™ Adenovirus Titer ELISA Kit, 192 assays
Supplier:  Cell Biolabs Inc VPK110
Encompass_Preferred
Traditional plaque forming unit assays (PFU) used to measure adenoviral functional titer are tedious and can take up to 10 days for results. Cell Biolabs' QuickTiter™ Adenovirus Titer ELISA Kit measures functional titer in less time (2.5 days vs. 10 days) with greater accuracy than traditional PFU assays. Each kit provides sufficient reagents for 192 assays.
Catalog No. NC9753057