Learn More
Meso Scale Discovery VPLEX Angiogenesis PNL 1HMNKit
SDP
Supplier:  Meso Scale Discovery K15190D4
Encompass
The Meso Scale Discovery V-PLEX Angiogenesis Panel 1 Human Kit is used to simultaneously measure VEGF VEGF-C VEGF-D and PlGF in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample consumption The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG Read Buffer Calibrator Diluent and lyophilized standards to generate a calibration curve This multiplex immunoassay allows angiogenesis biomarker profiling from a single sample in just 3 5 hours
Catalog No. 50-000-78910
Add to Cart
Request a Quote